DNA methylation kit, 5-hmC, BIOARRAY™
Leverandør: ENZO LIFE SCIENCES
ENZ450110050
ENZOENZ450110050EA 9600 NOK
ENZOENZ450110050
DNA methylation kit, 5-hmC, BIOARRAY™
Assay Nucleic Acid Assays
BIOARRAY™ 5-hmC Methylation Kit is used for the sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.
  • Single tube reaction format
  • Suitable for analysis by a variety of downstream applications
  • Flexible set-up for custom assay design
Leveringsinformasjon: Kit contains 5-hmC Glucosyltransferase (5-hmC), 10X 5-hmC reaction buffer, 10X UDPG (uridine diphosphoglucose), MspI restriction enzyme, 5-hmC control DNA, qPCR primer 1, qPCR primer 2 and 5-DNA clean up concentrator kit.