EpiXtract® total histone extraction kit
Leverandør: ENZO LIFE SCIENCES
ENZ450140100
ENZOENZ450140100EA 2975 NOK
ENZOENZ450140100
EpiXtract® total histone extraction kit
Proteinekstraksjons Protein Extraction Kits
Rapid, reliable method to isolate histone proteins in mammalian cells and tissues for histone modification applications.
For the extraction of total histone proteins from mammalian cells or tissues.
EpiXtract® Total Histone Extraction Kit provides a simple and reliable method to isolate histone proteins in one hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Bestillingsinformasjon: Contains 10x Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .