EpiXtract® Nuclear protein isolation kit
Leverandør: ENZO LIFE SCIENCES
ENZ450160100
ENZOENZ450160100EA 3675 NOK
ENZOENZ450160100
EpiXtract® Nuclear protein isolation kit
Proteinekstraksjons Protein Extraction Kits
Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues.
For the extraction of nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins.
Bestillingsinformasjon: Contains Pre-extraction Buffer, Extraction Buffer, Protease Inhibitor Cocktail, DTT.