Red hydrogen peroxide assay kit
Leverandør: ENZO LIFE SCIENCES
ENZ51004
ENZOENZ51004EA 5025 NOK
ENZOENZ51004
Red hydrogen peroxide assay kit
Assay Cellulær analyse
Red Hydrogen Peroxide Assay Kit provides a simple absorbance-based or fluorometric assay to detect and quantify hydrogen peroxide or peroxidase activity in biochemical assays, cell extracts and in live cells. It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit is an optimized ‘mix and read’ assay that is suitable for both 96- and 348-well formats and is compatible with HTS liquid handling.
  • Homogenous assay format, fully compatible with HTS automation
  • Detect as little as 10 picomoles of hydrogen peroxide
  • Mix and read reagents, with minimal hands-on time
Leveringsinformasjon: Kit includes Red Peroxidase Substrate (1 Vial), Horseradish Peroxidase (HRP) (20 units), Hydrogen Peroxide (H2O2) Stock Solution (200 µl), DMSO (1 ml), Assay Buffer (100 ml).