ROS-ID™ total ROS/superoxide detection kit
Leverandør: ENZO LIFE SCIENCES
ENZ51010
ENZOENZ51010EA 4375 NOK
ENZOENZ51010
ROS-ID™ total ROS/superoxide detection kit
Assay Cellulær analyse
The kit is designed to directly monitor real time reactive oxygen species (ROS) production and Specifically Superoxide in live cells using fluorescence microscopy or flow cytometry.
  • Distinguishes between different reactive species, such as hydrogen peroxide, peroxynitrite and hydroxyl radicals
  • High sensitivity, specificity and accuracy for live cell studies
  • Compatible with major components of tissue culture media (phenol red, FBS and BSA)
  • Complete set of reagents, including ROS inducers and scavengers
  • Validated on microscopy, flow cytometry, and microplates
Through the combination of these two specific fluorescent probes, the kit provides a simple and specific assay for the real-time measurement of global levels of reactive oxygen species (ROS), and specifically superoxide in living cells. The Total ROS/Superoxide Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in suspension) or 2 x 96 microplate assays.
Leveringsinformasjon: Kit contains Oxidative Stress Detection Reagent (Green), 300 nmoles, Superoxide Detection Reagent (Orange), 300 nmoles, ROS Inducer (Pyocyanin), 1 µmole, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and Wash Buffer Salts, 1 pack.