Fura 2-AM 95 HPLC_ASSAY_METHOD, ultrapure

Leverandør: ENZO LIFE SCIENCES

Synonymer: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

ENZ52006
ENZOENZ52006EA 2675 NOK
ENZOENZ52006
Fura 2-AM 95 HPLC_ASSAY_METHOD, ultrapure
Fura 2-AM

Calcium dye


Fura-2 acetomethoxy (AM) is one of the most widely used calcium indicators for homogenous quantitative dual wavelength ratiometric cell measurements. Fura-2 AM is particularly useful for digital imaging microscopy. It is less susceptible to photobleaching than Indo-1. The probe is excited only by UV light, which results in significantly less interference by visible wavelength excitable fluorescent compounds. One application of the probe is as a useful counter screen tool for GPCR and calcium channel hits identified using calcium indicators excited at 488 nm in primary screens. Fura-2 AM can be easily loaded into cells by passive incubation. Wavelength Maxima: Excitation 370 nm, Emission 476 nm.

Formel: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036976
CAS-nummer: 108964-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8479853873918dc948f6063302d1e3f6103ee7a3&vid=c3bc1d5b77dcf889948bc27fe4d1ff5d6dc4cd53&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR