Indo-1 AM ≥95% (by HPLC), Ultrapure

Leverandør: ENZO LIFE SCIENCES

Synonymer: 1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylester, Indo-1-(acetoxymethyl) ester, 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, Indo-1 AM Ester

ENZ52008
ENZOENZ52008EA 2725 NOK
ENZOENZ52008
Indo-1 AM ≥95% (by HPLC), Ultrapure
Indo-1 AM

Calcium dye


Indo-1 acetomethoxy (AM) is a popular UV-excitable calcium indicator, similar to Fura-2. In contrast to Fura-2, Indo-1 has a dual emission peak. The main emission peak in calcium-free solution is 475nm while in the presence of calcium the emission is shifted to 400 nm. Indo-1 AM is a membrane permeable probe and can be easily loaded into cells by passive incubation. The probe is widely used in flow cytometry. For this application, it is critical to have an instrument with an argon laser tuned to UV or a heliumcadmium laser available. Wavelength Maxima: Excitation 346 nm, Emission 475 nm.

Formel: C₄₇H₅₁N₃O₂₂
MW: 1009,91 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036955
CAS-nummer: 112926-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f5f1ab1b1d2cd22911daf3fac43fe2750d0eaf1&vid=82e9b619060ea198884fcc6ef531aba9fcff0663&prid=web&clid=SAPDAM
New Product

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR