Rhodamine 123 ≥95% (by HPLC), Ultrapure

Leverandør: ENZO LIFE SCIENCES

Synonymer: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester

ENZ52307
ENZOENZ52307EA 1950 NOK
ENZOENZ52307
Rhodamine 123 ≥95% (by HPLC), Ultrapure
Rhodamine 123

Mitochondria dye


Rhodamine 123 is a cell-permeant, cationic, green-fluorescent dye that is readily sequestered by active mitochondria without cytotoxic effects. Rhodamine 123 can be used in multi-parametric analysis, without fluorescence interference, in combination with common protein labeling dyes such as cyanine-5 and AMCA. With respect to apoptosis, the presence of mitochondrial membrane potential can be probed with rhodamine 123 while the structure and integrity of mitochondria can be assessed using 10-N-nonyl-acridine orange. Wavelength Maxima: Excitation 507 nm, Emission 529 nm.

Formel: C₂₁H₁₇ClN₂O₃
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00012664
CAS-nummer: 62669-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f698819dd5d857d7a3f34056f26359bafecfde9&vid=7fa2c58162af54173b3962ce3c11649ec305d922&prid=web&clid=SAPDAM
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR