Anti-NPC Mouse Monoclonal Antibody [clone: MAb414]

Leverandør: ENZO LIFE SCIENCES
ENZABS1050100
ENZOENZABS1050100EA 6025 NOK
ENZOENZABS1050100
Anti-NPC Mouse Monoclonal Antibody [clone: MAb414]
Antistoffer
Recommended Applications: ICC, IHC, IP, WB, Immunoelectron microscopy

Type: Primary
Antigen: NPC
Clonality: Monoclonal
Clone: MAb414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast, Xenopus, Vertebrates
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR