Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]

Leverandør: ENZO LIFE SCIENCES
ENZABS1220100
ENZOENZABS1220100EA 7775 NOK
ENZOENZABS1220100
Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]
Antistoffer
Recommended Applications: Flow Cytometry, IF, IHC

Species reactivity: Mouse

Type: Primary
Antigen: CD205
Clonality: Monoclonal
Clone: NLDC-145
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol CD205
Environmentally Preferable
Antigen synonymer CD205|Lymphocyte antigen 75
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon NLDC-145
Reaktivitet Mouse
Vert Rat
Isotype IgG2a
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Immunogen Mouse lymphoid tissue stroma.
Skjema Liquid
Rensing Purified using QMA Hyper D ion exchange chromatography
Lagringsbuffer In TRIS-NaCl, pH 8.
Lagringstemp −80 °C
Frakttemp. Shipped on Blue Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR