Anti-CD80 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: mAb 104]

Leverandør: ENZO LIFE SCIENCES
ENZABS1510100
ENZOENZABS1510100EA 9000 NOK
ENZOENZABS1510100
Anti-CD80 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: mAb 104]
Antistoffer
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: mAb 104
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR