Anti-CD38 Mouse Monoclonal Antibody (Biotin) [clone: HB7]

Leverandør: ENZO LIFE SCIENCES
ENZABS1600100
ENZOENZABS1600100EA 7575 NOK
ENZOENZABS1600100
Anti-CD38 Mouse Monoclonal Antibody (Biotin) [clone: HB7]
Antistoffer
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR