Anti-IgE Mouse Monoclonal Antibody (Biotin) [clone: I27]

Leverandør: ENZO LIFE SCIENCES
ENZABS2250100
ENZOENZABS2250100EA 7925 NOK
ENZOENZABS2250100
Anti-IgE Mouse Monoclonal Antibody (Biotin) [clone: I27]
Antistoffer
Recommended Applications: Flow Cytometry, WB

Species reactivity: Human

Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: I27
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammalian
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol IgE
Environmentally Preferable
Antistofftype Secondary
Klonalitet Monoclonal
Konjugering Biotin
Klon I27
Reaktivitet
Vert Mouse
Isotype IgG1
Western blot Yes
Flowcytometri Yes
Immunogen Purified human IgE (preincubated with mouse serum anti human IgG).
Skjema Liquid
Lagringsbuffer In PBS containing 50% glycerol
Lagringstemp −80 °C
Frakttemp. Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR