DNA ladder 50 to 1500 bp, AMPIGENE®
Leverandør: ENZO LIFE SCIENCES
ENZGEN1020100
ENZOENZGEN1020100EA 1600 NOK
ENZOENZGEN1020100
DNA ladder 50 to 1500 bp, AMPIGENE®
Elektroforese reagenser Molekylvekt markører
AMPIGENE® DNA ladder contains DNA fragments ranging from 50 bp to 1500 bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 200 bp, 500 bp and 1500 bpto serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5 µl load, for determining the approximate mass of DNA in samples with comparable intensity and sizes.
  • Ready to use format can be loaded straight onto a gel
  • Evenly spaced bands with intensified reference bands
  • Stable at room temperature
Leveringsinformasjon: Kit contains ready to use AMPIGENE® DNA ladder 50 – 1500 bp and sample loading buffer.