Amp'd® HSP70 high sensitivity ELISA Kit
Leverandør: ENZO LIFE SCIENCES
ENZKIT1010001
ENZOENZKIT1010001EA 10325 NOK
ENZOENZKIT1010001
Amp'd® HSP70 high sensitivity ELISA Kit
Assay ELISAs
Ultra-sensitive (7 pg/ml)Amp'd® HSP70 high sensitivity ELISA kit enabling the ability to use less sample and detect both baseline and upregulated levels of human, mouse and rat Hsp70 (Hsp72), a major chaperone, cancer biomarker, and key cell stress regulator.
For the quantitative determination of normal and upregulated (stressed) levels of HSP70 in serum and plasma of human, mouse, and rat origin.
The Amp'd® HSP70 high sensitivity ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Assay Buffer 28, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and Amp'd (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål HSP70
Primary antibody reactivity
Vert
Sensitivitet 7 pg/ml (range 39 - 500 pg/ml)