AMPIGENE® 1-Step RT-PCR kit
Leverandør: ENZO LIFE SCIENCES
ENZKIT1050050 ENZKIT1050200 ENZKIT1051000
ENZOENZKIT1050050EA 3125 NOK
ENZOENZKIT1050050 ENZOENZKIT1050200 ENZOENZKIT1051000
AMPIGENE® 1-Step RT-PCR kit
Nukleinsyre reagenser RT-PCR and cDNA Reagents
AMPIGENE® 1-Step RT-PCR Kit for optimised and efficient cDNA synthesis and PCR in a single tube.
AMPIGENE® 1-Step RT-PCR Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and PCR in a single tube.The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity.
Bestillingsinformasjon: Contains 2x 1-Step Mix (AMPIGENE® HS Taq Polymerase, 6mM MgCl2, 2mM dNTPs, enhancer, and stabilizers), 20x RTase (with RNase inhibitor).