Cellular senescence live cell analysis assay
Leverandør: ENZO LIFE SCIENCES
ENZKIT1300010
ENZOENZKIT1300010EA 5225 NOK
ENZOENZKIT1300010
Cellular senescence live cell analysis assay
Assay Cellulær analyse
Simple assay to quantify senescence-associated β-galactosidase activity, in live cells, using flow cytometry or fluorescent microscopy.
For the study of cellular senescence in live cells.
Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells.
Bestillingsinformasjon: Contains Cell pretreatment solution (1000x) and SA-?-gal Substrate (200x).