TGF-α ELISA kit, Enzo Life Sciences
Leverandør: ENZO LIFE SCIENCES
ENZKIT1320001
ENZOENZKIT1320001EA 9625 NOK
ENZOENZKIT1320001
TGF-α ELISA kit, Enzo Life Sciences
Assay ELISAs
Highly sensitive ELISA kit enabling detection of TGF-α in 3,5 hours.
  • Sensitive measurement of TGF-α, detecting as little as 9,375 pg/ml
  • Negligible reactivity from similar growth factors
  • High throughput format with results in 3,5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TGF-α in serum, plasma, cell culture supernatant, tissue homogenates in human samples.
The TGF-α ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Standard, Detection Antibody, Detection Antibody Diluent, Sample Diluent, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.