IL-5 ELISA kit, grow cells
Leverandør: ENZO LIFE SCIENCES
ENZKIT1390001
ENZOENZKIT1390001EA 11050 NOK
ENZOENZKIT1390001
IL-5 ELISA kit, grow cells
Assay ELISAs
Highly sensitive IL-5 ELISA kit enabling detection of IL-5 in serum, plasma, urine and cell culture supernates in just 3 hours.
  • Sensitive measurement of IL-5, detecting as little as 2 pg/ml.
  • Negligible reactivity to related proteins.
  • High throughput with results in three hours.
  • Fully quantitative results that pass semi-quantitative Western blot analysis.
For the quantitative determination of IL-5 in cell culture supernates, serum, plasma (Heparin & EDTA), urine, in human samples.
The IL-5 ELISA kit is a colorimetric sandwich immunoassay kit with results in three hours.
Bestillingsinformasjon: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Substrate, and Stop Solution.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål IL-5
Primary antibody reactivity
Vert
Sensitivitet <2 pg/ml (7,8 - 500 pg/ml)