Transferrin ELISA kit, Enzo Life Sciences
Leverandør: ENZO LIFE SCIENCES
ENZKIT1430001
ENZOENZKIT1430001EA 8875 NOK
ENZOENZKIT1430001
Transferrin ELISA kit, Enzo Life Sciences
Assay ELISAs
Highly sensitive Transferrin ELISA kit enabling detection of transferrin in serum and plasma in just 2 hours.
  • Sensitive measurement of transferrin, detecting as little as 4,6 ng/ml
  • Negligible cross reactivity with similar proteins
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of transferrin in human serum, human plasma and canine serum. The Transferrin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Tracer, Conjugate, Substrate, and Stop Solution.