BNP fragment ELISA kit, grow cells
Leverandør: ENZO LIFE SCIENCES
ENZKIT1520001
ENZOENZKIT1520001EA 8975 NOK
ENZOENZKIT1520001
BNP fragment ELISA kit, grow cells
Assay ELISAs
Sensitive BNP fragment ELISA kit enabling detection of BNP fragment in serum and plasma.
  • Quantitative determination of human BNP fragment
  • Colorimetric, competitive enzyme immunoassay
For the quantitative determination of human BNP fragment in serum and plasma. The BNP Fragment ELISA kit is a colorimetric competitive enzyme immunoassay kit.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Primary antibody reactivity
Analysetype Competitive
Vert
Sensitivitet 171 pmol/L (0-6400 pmol/L)