AMPIGENE® qPCR 1-Step Probe Kit No-ROX
Leverandør: ENZO LIFE SCIENCES
ENZNUC1120200 ENZNUC1121000
ENZOENZNUC1120200EA 4350 NOK
ENZOENZNUC1120200 ENZOENZNUC1121000
AMPIGENE® qPCR 1-Step Probe Kit No-ROX
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
Optimised and efficient cDNA synthesis and probe-based qPCR in a single tube.
AMPIGENE® qPCR 1-Step Probe Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE® qPCR 1-Step Probe Kits are designed for use on a variety of probes including: Taqma ®, Molecular Beacons®, and Scorpion probes®.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR 1-Step Probe Kits come with either Lo-ROX, Hi-ROX, or No-ROX concentration. For Lo-ROX and Hi-ROX, see Prod. Nos. ENZ-NUC110 and ENZ-NUC111. Choose the appropriate ROX concentration for the instrument.
Bestillingsinformasjon: Contains 2x AMPIGENE® qPCR 1-Step Probe Mix No-ROX (includes dNTP, and polymerases) 20x RTase (with RNase inhibitor).
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon qPCR probe
One step Ja