Mouse recombinant IL-1beta
Leverandør: ENZO LIFE SCIENCES
ENZPRT1360010
ENZOENZPRT1360010EA 3600 NOK
ENZOENZPRT1360010
Mouse recombinant IL-1beta
Proteiner
Produced in E.coli.
Bestill nå
SPESIFIKASJONER
Protein/peptide name IL-1beta
Environmentally Preferable
Protein synonyms Catabolin|Interleukin-1β
Protein/peptide type Recombinant
Arter Mouse
Kilde
Renhet >98% (SDS-PAGE gel & HPLC)
Formulering Lyophilized from 10 mm sodium citrate, pH 4,0
Molekylvekt ~17.5 kDa