Human recombinant GDNF

Leverandør: ENZO LIFE SCIENCES

ENZPRT1450010
ENZOENZPRT1450010EA 3600 NOK
ENZOENZPRT1450010
Human recombinant GDNF
Proteiner

Produced in E.coli.

Bestill nå

SPESIFIKASJONER

Protein/peptide name GDNF
Protein synonyms Glial cell-derived neurotrophic factor|ATF|Astrocyte-derived trophic factor|hGDNF
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde
Renhet >98% (SDS-PAGE gel & HPLC)
Formulering Lyophilised from 10 mm sodium citrate, containing 150 mm NaCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR