Human Fibrinogen (from plasma)
Leverandør: ENZO LIFE SCIENCES
ENZPRT1570100
ENZOENZPRT1570100EA 1950 NOK
ENZOENZPRT1570100
Human Fibrinogen (from plasma)
Proteiner
Isolated from human plasma. Plasminogen depleted.
Bestill nå
SPESIFIKASJONER
Protein/peptide name Fibrinogen
Protein synonyms thromboembolism|Factor 1|cardiovascular Disease|atherosclerotic lesion formation|blood coagulation system|Atherosclerosis|Stroke|ischaemic heart disease|IHD|cardiovascular
Environmentally Preferable
Arter Human
Kilde Plasma
Renhet >95%
Formulering Liquid. In 20mm sodium citrate-HCl, pH 7,4
Molekylvekt ~330 kDa