T7 RNA polymerase
Leverandør: Thermo Fisher Scientific
EP0111
EP0111EA 440 NOK
EP0111
T7 RNA polymerase
Nukleinsyre reagenser RNA Synthesis and Transcription
T7 RNA polymerase is ideal for synthesis of unlabeled and labeled RNA. It catalyses the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter. It is also used in the study of RNA secondary structure and RNA-protein interactions, RNA splicing.