Termocycler, Mastercycler® ep
Leverandør: EPPENDORF
5345000.013 5341000.043 5341000.540 5344000.044 5344000.540 5349810.001
732-0124EA 0 NOK
732-0124 732-0909 732-0411 732-0915 732-0413 EPPE5349810.001
Termocycler, Mastercycler® ep
PCR-maskiner
The Mastercycler® ep family of thermal cyclers are distinguished by having thermoblocks with extremely fast heat-ing and cooling speeds, and unique heated lid technology (ESP - Electronic Sample Protection), which avoids the preheating of samples and minimises unspecific annealing. The gradient option of each Mastercycler® ep system features SteadySlope® technology to ensure that the heating and cooling ramp rates are identical in both gradient and normal operation. Programming with the control panel is easy and intuitive, allowing novice users to get started quickly and safely.