Thermal cyclers, Mastercycler® nexus flat
Leverandør: EPPENDORF
6330000.013
EPPE6330000.013EA 0 NOK
EPPE6330000.013
Thermal cyclers, Mastercycler® nexus flat
PCR-maskiner
Special applications require special consumables. With its flat block and no wells, the Mastercycler® nexus flat offers the optimal foundation for slides and other unconventional consumable formats. When doing in situ PCR, results are normally influenced by the characteristic temperature transfer of the corresponding in situ adapter. The Mastercycler® nexus flat can heat and cool slides directly without the need for an adapter.
  • Flat block without wells
  • Intuitive graphic programming
  • Small footprint
  • Up to two more units can be connected to the central unit
  • E-