Mastercycler® nexus X2 PCR Thermal Cycler

Leverandør: EPPENDORF
6336000.040 6337000.043 6337000.019 6336000.015 6338000.012 6339000.016
731-0431EA 0 NOK
731-0431 EPPE6337000.043 EPPE6337000.019 732-3666 EPPE6338000.012 EPPE6339000.016
Mastercycler® nexus X2 PCR Thermal Cycler
PCR-maskiner
For a high PCR workload, the Eppendorf® Mastercycler nexus X2 offers efficiency with two blocks (32-well and 64-well), allowing simultaneous runs of two independent protocols. Accommodates a range of consumables up to 0,5 ml tubes. For increased capacity, connect with up to three additional units.

  • Asymmetrically divided block for running different assays in parallel, one 32-well and one 64-well block
  • Intuitive graphic programming
  • Universal block for strips, 0,1, 0,2 and 0,5 ml PCR tubes and fits for divisible PCR plates
  • Small footprint
  • Optional gradient
More Product Information
Bestill nå

SPESIFIKASJONER

Modell Mastercycler® nexus X2 Mastercycler® nexus X2 Mastercycler® nexus GX2 Mastercycler® nexus GX2e Mastercycler® nexus X2e
Blokkhomogenitet ±0,3 °C at 20...72 °C
±0,4 °C at 95 °C
±0,3 °C at 20...72 °C
±0,4 °C at 95 °C
±0,3 °C at 20...72 °C
±0,4 °C at 95 °C
±0,3 °C at 20...72 °C
±0,4 °C at 95 °C
±0,3 °C at 20...72 °C
±0,4 °C at 95 °C
Environmentally Preferable
Blokkmaterial Aluminum Aluminum Aluminum Aluminum Aluminum
Kontrollnøyaktighet ± 0,2 °C ± 0,2 °C ± 0,2 °C ± 0,2 °C ± 0,2 °C
Nedkjølingshastighet max. 2 °C/s max. 2 °C/s max. 2 °C/s max. 2 °C/s max. 2 °C/s
Gradient - - + + -
Spennvidde for gradient (maks./min.) 1...12 °C 1...12 °C
Temperaturområde for gradient 30...99 °C 30...99 °C
Oppvarmingsområde max. 3 °C/s max. 3 °C/s max. 3 °C/s max. 3 °C/s max. 3 °C/s
Grensesnitt USB, Ethernet, CAN in, CAN out USB, Ethernet, CAN in, CAN out USB, Ethernet, CAN in, CAN out USB, Ethernet, CAN in, CAN out Ethernet, CAN in, CAN out
Lid flexlid® technology, with TSP (lid descent) flexlid® technology, with TSP (lid descent) flexlid® technology, with TSP (lid descent) flexlid® technology, with TSP (lid descent) flexlid® technology, with TSP (lid descent)
Temperaturområde for lokk 37...110 °C 37...110 °C 37...110 °C 37...110 °C 37...110 °C
Prøvekapasitet 64/32×0,2 ml PCR tubes or up to 24/10×0,5 ml PCR tubes 64/32×0,2 ml PCR tubes or up to 24/10×0,5 ml PCR tubes 64/32×0,2 ml PCR tubes or up to 24/10×0,5 ml PCR tubes 64/32×0,2 ml PCR tubes or up to 24/10×0,5 ml PCR tubes 64/32×0,2 ml PCR tubes or up to 24/10×0,5 ml PCR tubes
Temperaturområde 4...99 °C (temp. control range of the block) 4...99 °C (temp. control range of the block) 4...99 °C (temp. control range of the block) 4...99 °C (temp. control range of the block) 4...99 °C (temp. control range of the block)
Uniformitet temperatur ≤ ±0.4 °C ≤ ±0.4 °C ≤ ±0.4 °C ≤ ±0.4 °C ≤ ±0.4 °C
B×D×H 250×412×321 mm 250×412×330 mm 250×412×321 mm 250×412×321 mm 250×412×321 mm
Vekt 11 kg 11 kg 11 kg 11 kg 11 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR