Thermal cyclers, Mastercycler® nexus X1
Leverandør: EPPENDORF
6348000.010
EPPE6348000.010EA 0 NOK
EPPE6348000.010
Thermal cyclers, Mastercycler® nexus X1
PCR-maskiner
The Mastercycler® nexus X1 combines the modern and intuitive software from the Mastercycler® nexus with a fast 96-well silver block for increased heating and cooling rates.
  • Heating rate: 5 °C/s
  • Fast silver block
  • Flexlid™ concept allows use of different consumables with automatic height adjustment of the lid
  • Intuitive graphic programming
  • Small footprint
  • Up to two other units can be connected to a central unit
  • E-mail notification
*To run a Mastercycler nexus® with the suffix 'e', a Mastercycler® nexus model without such a suffix is needed. Up to 2 unitsMore Product Information