Mastercycler® X40 PCR Thermal Cycler
Leverandør: EPPENDORF
6381000.018
EPPE6381000.018EA 0 NOK
EPPE6381000.018
Mastercycler® X40 PCR Thermal Cycler
PCR-maskiner
Mastercycler X40 has classic features such as a 12-column gradient and excellent block temperature homogeneity that are enhanced by an easy, intuitive user interface, a touchscreen and a protective SafeLid to protect your samples from evaporation.
  • 12-column gradient
  • SafeLid for evaporation protection
  • Small footprint (less than a piece of paper)
  • Lightweight
  • Touchscreen
  • Intuitive software and user interface
  • Two-year warranty
  • Program migration feature
  • Visionize Labsuite connection possible
  • Low noise level
  • VisioNize® touch enabled remote device monitoring and