Flasker, vid munn, med skrugjenger, uten skrulokk
Leverandør: EP SCIENTIFIC
012-0120BP 012-0250BP 012-0500BP
EPSC012-0120BPEA 7375 NOK
EPSC012-0120BP EPSC012-0250BP EPSC012-0500BP
Flasker, vid munn, med skrugjenger, uten skrulokk
Flasker Plastflasker
HDPE, translucent.
  • Without screw caps
  • Wide mouth for easy filling and sample retrieval
  • Recommended for general purpose sampling where cleaning and certification is not required
  • Economically bulk packed without caps
Wide-mouth packers are general-purpose containers, bulk and unassembled. Unprocessed, ready for own cleaning procedure and assembly with screw caps purchased separately.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement