Flasker, vannprøver, koliforme, steril
Leverandør: EP SCIENTIFIC
156-120ST
EPSC156-120STEA 10150 NOK
EPSC156-120ST
Flasker, vannprøver, koliforme, steril
Flasker Vannprøveflasker
PP, PE or PS.
  • Certified sterile; manufactured under an ISO certified quality management system
  • Threaded screw closure for easy aseptic sample collection and secure cap engagement
  • Non-autofluorescent
  • Available pre-charged with a sodium thiosulfate (Na₂S₂O₃) tablet for sample de-chlorination, or without a tablet for un-chlorinated water sources
  • 100 ml fill line
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.