Water sampling bottles, glass, wide neck, amber
Leverandør: EP SCIENTIFIC
341-0120 341-0250 341-0500 341-0950 341-2500 341-1250 141-0060 141-0120 141-0250 141-2500 141-0500 141-0950 341-0060 241-0060 241-0120 241-0250 241-0500 241-0950
EPSC341-0120EA 1350 NOK
EPSC341-0120 EPSC341-0250 EPSC341-0500 EPSC341-0950 EPSC341-2500 EPSC341-1250 EPSC141-0060 EPSC141-0120 EPSC141-0250 EPSC141-2500 EPSC141-0500 EPSC141-0950 EPSC341-0060 EPSC241-0060 EPSC241-0120 EPSC241-0250 EPSC241-0500 EPSC241-0950
Water sampling bottles, glass, wide neck, amber
Flasker Vannprøveflasker
Type III soda-lime glass, amber, with screw cap
  • With PTFE-lined white PP closed top cap (assembled on bottles)
  • Certified to meet or exceed EPA Performance Based Specifications for semi-volatile organics, pesticides, PCBs and metals analyses
  • Each jar individually barcoded with lot number and unique container number
These bottles are designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required. These bottles are processed the same way as the certified jars butMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement