Bottles, narrow neck, amber glass, Boston type with caps
Leverandør: EP SCIENTIFIC
145-4000 145-2360 345-2360 345-4000 245-4000 245-2360
EPSC145-4000EA 2125 NOK
EPSC145-4000 EPSC145-2360 EPSC345-2360 EPSC345-4000 EPSC245-4000 EPSC245-2360
Bottles, narrow neck, amber glass, Boston type with caps
Flasker Vannprøveflasker
Type III soda-lime amber glass, with screw cap.
  • Excellent chemical resistance
  • Good temperature resistance
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Sertifiseringer: Certified to meet EPA performance based specifications for semi-volatile organics, pesticides, PCBs and metals analyses. Available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses, Processed the same way as the certified jugs butMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement