Water sampling bottles, wide neck, short-profile amber
Leverandør: EP SCIENTIFIC
140-0120 140-0060 340-0060 340-0120 340-0250 140-0250 140-0250NC 240-0120 240-0060
EPSC140-0120EA 1400 NOK
EPSC140-0120 EPSC140-0060 EPSC340-0060 EPSC340-0120 EPSC340-0250 EPSC140-0250 EPSC140-0250NC EPSC240-0120 EPSC240-0060
Water sampling bottles, wide neck, short-profile amber
Flasker Vannprøveflasker
These jars, made of Type III soda-lime glass, amber, with white LDPE-lined PP screw cap, is designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is required.
  • Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses
  • Processed the same way as the certified jars but without certification documentation
  • Unprocessed and ready for your own cleaning procedure
  • Closures assembled on bottles
Leveringsinformasjon: CertifiedMore Product Information