Closed caps, PTFE lined
Leverandør: EP SCIENTIFIC
B267-2414UBB
EPSCB267-2414UBBEA 1650 NOK
EPSCB267-2414UBB
Closed caps, PTFE lined
Lokk Flaskelokk
These PTFE-lined closed caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement