Sample vials, closed-top
Leverandør: EP SCIENTIFIC
C126-0020 C136-0040 C146-0040 C226-0020 C236-0040 C246-0040 C326-0020 C336-0040 C346-0040
EPSCC126-0020EA 2400 NOK
EPSCC126-0020 EPSCC136-0040 EPSCC146-0040 EPSCC226-0020 EPSCC236-0040 EPSCC246-0040 EPSCC326-0020 EPSCC336-0040 EPSCC346-0040
Sample vials, closed-top
Vialer
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With closed-top white PP cap bonded with PTFE-lined silicone septum for secure sealing
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CoA is not included.