Dust covers
Leverandør: EP SCIENTIFIC
DC-VOA
EPSCDC-VOAEA 3375 NOK
EPSCDC-VOA
Dust covers
Lokk Lokk til vials
These PE dust covers are designed to fit all standard size VOA vial caps. They prevent contamination and protect critical samples during transport.
  • Allow for septum expansion
  • Pre cleaned dust covers available upon request