EPA vials with PP cap
Leverandør: EP SCIENTIFIC
139-20C 140-40C 139-20A 141-40A/CT 139-20C/CT 140-40C/CT S246-0040 S246-0060 139-20A/EP 139-20C/EP 140-40C/DB 140-40C/EP 140-40C/TS 139-20A/CT 141-40A/DB 141-40A/EP 141-40A/TS 141-60A 341-40A 341-60A GVB-100A
EPSC139-20CEA 5200 NOK
EPSC139-20C EPSC140-40C EPSC139-20A EPSC141-40A/CT EPSC139-20C/CT EPSC140-40C/CT EPSCS246-0040 EPSCS246-0060 EPSC139-20A/EP EPSC139-20C/EP EPSC140-40C/DB EPSC140-40C/EP EPSC140-40C/TS EPSC139-20A/CT EPSC141-40A/DB EPSC141-40A/EP EPSC141-40A/TS EPSC141-60A EPSC341-40A EPSC341-60A EPSCGVB-100A
EPA vials with PP cap
Vialer
The EPA vials are ideal for water sample collection for volatile organic analysis. The vials with unbonded septa have a molded-in retainer ring to keep septa firmly in place while allowing septa to be removed and replaced as needed. Also there are vials with the septum permanently bonded to the cap for secure sealing.
  • Used for water sample collection for volatile organic analysis (VOA)
  • Amber vials protect light-sensitive analytes from break-down
  • Open-top white PP cap with standard 3,18 mm thick silicone lined PTFE septum
  • Directly compatible with automation