Wipes, pre-saturated
Leverandør: ESSENTRA POROUS TECHNOLOGIES
6-LS7030-PC-24
EPTA6-LS7030-PC-24EA 6650 NOK
EPTA6-LS7030-PC-24
Wipes, pre-saturated
Papirservietter Tørkepapir og våtservietter Våtservietter
These polyester/cellulose wipes are pre-saturated with 70% IPA and 30% DIW. They are ideal for general and maintenance applications, as well as for industrial cleaning.
  • Pre-wetted with alcohol
  • Resealable pouch for easy dispensing
  • Excellent as a degreaser
  • Made in the USA
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904