Wipes, pre-saturated
Leverandør: ESSENTRA POROUS TECHNOLOGIES
6-LS964-685
EPTA6-LS964-685EA 5950 NOK
EPTA6-LS964-685
Wipes, pre-saturated
Papirservietter Tørkepapir og våtservietter Våtservietter
These polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
  • Perforated roll pre-wetted with alcohol
  • Canister with spring loaded lid prevents dying out and facilities dispensing
  • Alcohol saturates and evaporates quickly
  • Made in the USA
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904