Restriction enzymes, Restriction enzyme, Eco147I (StuI), Fermentas
Leverandør: Thermo Fisher Scientific
ER0421
ER0421EA 1125 NOK
ER0421
Restriction enzymes, Restriction enzyme, Eco147I (StuI), Fermentas
Nukleinsyre reagenser Restriction Enzymes
Thermo Scientific Eco147I (StuI) restriction enzyme recognises AGG^CCT sites and cuts best at 37 °C in B buffer (Isoschizomers: AatI, PceI, SseBI, StuI).
Applications include: Molecular cloning, restriction site mapping, genotyping, Southern blotting, restriction fragment length polymorphism (RFLP), SNP.
Kit is supplied as 3 vials: Eco1471 1000 units; 1 ml of 10X Buffer B; 1 ml of 10X Buffer Tango.