Restriction enzymes, PfoI, Fermentas
Leverandør: Thermo Fisher Scientific
ER1751
ER1751EA 2575 NOK
ER1751
Restriction enzymes, PfoI, Fermentas
Nukleinsyre reagenser Restriction Enzymes
PfoI restriction enzyme recognises T^CCNGGA sites and cuts best at 37 °C in Tango buffer.
  • Stringent quality control and industry leading manufacturing process
  • Colour-coded five buffer system
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Its application includes molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP)and SNP.
PdoI is CpG methylation-sensitive, dam methylation-sensitive, dcm methylation-sensitive and also sensitive to heat inactivation.
Leveringsinformasjon: It includes universal Tango buffer for double-digestions.