Ergonomic floor mats, ESD, Ergomat®
Leverandør: ERGOMAT
EFB-LIGHTGREY
ERGOEFB-LIGHTGREYEA 2200 NOK
ERGOEFB-LIGHTGREY
Ergonomic floor mats, ESD, Ergomat®
Gulvmatter Gulvmatter, støtdempende
The ESD mats Ergomat® EFB and EFS Complete are made from pure polyurethane and are designed to increase comfort while reducing trip hazards and body pain. These mats are ideal for areas that combine sitting and standing or where static electricity control is a must. The smooth version is ideal for areas where trolleys and carts are used.
  • Choice of smooth or bubbled upper surface
  • Fire retardant, resists moderate oils and liquids
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantees durability and ergonomic benefits