Ductless filtering fume hoods, Captair® Smart 391
Leverandør: ERLAB CAPTAIRE
AA616A00011A21
ERLAAA616A00011A21EA 65650 NOK
ERLAAA616A00011A21
Ductless filtering fume hoods, Captair® Smart 391
Avtrekksskap og kabinetter Avtrekkshetter
The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood. A simple light pattern will indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://no.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=17327607
  • The Erlab's unique Flex Filtration can manage most laboratory applications
  • New