Ductless filtering fume hoods, Captair® Smart 392
Leverandør: ERLAB CAPTAIRE
AA636A00011A21
ERLAAA636A00011A21EA 0 NOK
ERLAAA636A00011A21
Ductless filtering fume hoods, Captair® Smart 392
Avtrekksskap og kabinetter Avtrekkshetter
The Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://no.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=20532258
  • A simple light pattern and sound pulses indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough
  • Felxibility: Modular filtration column adapting to application changes