Stand for mobilux® LED
Leverandør: ESCHENBACH OPTIK
1511003
ESCH1511003EA 450 NOK
ESCH1511003
Stand for mobilux® LED
Stativer
High quality stand for the mobilux® LED that can be used in the flat position for tremble-free reading and viewing, as well as in an upright position to free hands for carrying out work behind the lens.
  • Leaves both hands free for reading and working
  • Easy to use
  • Maintains correct distance of use
The stand is equipped with a 1/4" thread. This enables it to be screwed on to a standard flexible tripod. The stand enables to freely adjust the distance of use and it can be used with all mobilux® LED magnifiers.