Conductivity electrode, Eutech
Leverandør: EUTECH
93X546101
EUTE93X546101EA 3975 NOK
EUTE93X546101
Conductivity electrode, Eutech
Elektroder Konduktivitetselektrode
Conductivity/temperature sensor with epoxy body and stainless steel rings. For use with the following Eutech meters: CON 700 / PC 700 / CON 2700 / PC 2700.