ORP electrode, Eutech
Leverandør: EUTECH
ECFC7960101B
EUTEECFC7960101BEA 2425 NOK
EUTEECFC7960101B
ORP electrode, Eutech
Elektroder Redox Electrodes
General purpose plastic-body ORP electrode for use with all pH meters with BNC input connector.